Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGATAGGAAGTAAGATGCATCAAG[C/G]AAAATTGGAAAAACGTGGTTTTCCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605290 | |||||||||||||||||||||||
Literature Links: |
OPA1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
OPA1 - OPA1, mitochondrial dynamin like GTPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015560.2 | Intron | NP_056375.2 | ||||
NM_130831.2 | Intron | NP_570844.1 | ||||
NM_130832.2 | Intron | NP_570845.1 | ||||
NM_130833.2 | Intron | NP_570846.1 | ||||
NM_130834.2 | Intron | NP_570847.2 | ||||
NM_130835.2 | Intron | NP_570848.1 | ||||
NM_130836.2 | Intron | NP_570849.2 | ||||
NM_130837.2 | Intron | NP_570850.2 | ||||
XM_017006500.1 | Intron | XP_016861989.1 |