Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTACATCTTCCAGATGCTGGTC[C/T]GTCCCAGCCTGCCTCTGCTGGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606551 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LZTS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LZTS1 - leucine zipper tumor suppressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021020.3 | 5548 | UTR 3 | NP_066300.1 | |||
XM_005273394.3 | 5548 | UTR 3 | XP_005273451.1 | |||
XM_011544383.2 | 5548 | UTR 3 | XP_011542685.1 | |||
XM_011544384.2 | 5548 | UTR 3 | XP_011542686.1 | |||
XM_011544385.2 | 5548 | UTR 3 | XP_011542687.1 | |||
XM_011544386.2 | 5548 | UTR 3 | XP_011542688.1 |