Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATATCCAAAGGGTATAGTTAACC[A/G]TTATCCAGAGGATTGAAAGAATAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603517 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BCL10 - B-cell CLL/lymphoma 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320715.1 | Intron | NP_001307644.1 | ||||
NM_003921.4 | Intron | NP_003912.1 | ||||
XM_011542397.2 | Intron | XP_011540699.1 | ||||
XM_011542398.2 | Intron | XP_011540700.1 | ||||
XM_011542399.2 | Intron | XP_011540701.1 |
C1orf52 - chromosome 1 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC646626 - uncharacterized LOC646626 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |