Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGTTTTGTGGGGTTCGCTGCCC[A/G]AGCTGCTGGTCAGATTTGTTGGGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607033 MIM: 604709 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFB1M PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TFB1M - transcription factor B1, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016020.3 | 2109 | Intron | NP_057104.2 | |||
XM_005267006.4 | 2109 | Intron | XP_005267063.1 | |||
XM_011535870.1 | 2109 | Intron | XP_011534172.1 | |||
XM_011535871.1 | 2109 | Intron | XP_011534173.1 | |||
XM_011535872.2 | 2109 | UTR 3 | XP_011534174.1 | |||
XM_011535873.2 | 2109 | Intron | XP_011534175.1 | |||
XM_017010917.1 | 2109 | Intron | XP_016866406.1 |
TIAM2 - T-cell lymphoma invasion and metastasis 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010927.2 | 2109 | Intron | NP_001010927.1 | |||
NM_012454.3 | 2109 | Intron | NP_036586.2 |
Set Membership: |
HapMap |