Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614556 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARID1B - AT-rich interaction domain 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017519.2 | Intron | NP_059989.2 | ||||
NM_020732.3 | Intron | NP_065783.3 | ||||
XM_005267069.4 | Intron | XP_005267126.2 | ||||
XM_011535984.2 | Intron | XP_011534286.2 | ||||
XM_011535988.2 | Intron | XP_011534290.1 | ||||
XM_017011103.1 | Intron | XP_016866592.1 | ||||
XM_017011104.1 | Intron | XP_016866593.1 | ||||
XM_017011105.1 | Intron | XP_016866594.1 | ||||
XM_017011106.1 | Intron | XP_016866595.1 | ||||
XM_017011107.1 | Intron | XP_016866596.1 | ||||
XM_017011108.1 | Intron | XP_016866597.1 | ||||
XM_017011109.1 | Intron | XP_016866598.1 |
MIR4466 - microRNA 4466 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |