Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTGGGAAGGTGAGCAGCACACT[C/T]ATTTAATCCAAGTTTCTTTTCCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERICH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ERICH3 - glutamate rich 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002912.4 | 5714 | UTR 3 | NP_001002912.4 | |||
XM_017000274.1 | 5714 | UTR 3 | XP_016855763.1 | |||
XM_017000275.1 | 5714 | Intron | XP_016855764.1 | |||
XM_017000276.1 | 5714 | UTR 3 | XP_016855765.1 | |||
XM_017000277.1 | 5714 | UTR 3 | XP_016855766.1 | |||
XM_017000278.1 | 5714 | UTR 3 | XP_016855767.1 | |||
XM_017000279.1 | 5714 | UTR 3 | XP_016855768.1 | |||
XM_017000280.1 | 5714 | UTR 3 | XP_016855769.1 | |||
XM_017000281.1 | 5714 | UTR 3 | XP_016855770.1 | |||
XM_017000282.1 | 5714 | UTR 3 | XP_016855771.1 | |||
XM_017000283.1 | 5714 | UTR 3 | XP_016855772.1 | |||
XM_017000284.1 | 5714 | UTR 3 | XP_016855773.1 | |||
XM_017000285.1 | 5714 | UTR 3 | XP_016855774.1 |
ERICH3-AS1 - ERICH3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |