Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGATGAACTCGGTAGCTCTGGA[C/G]ATCCTGTCTTGACCTCCTCATCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LRRC20 - leucine rich repeat containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278211.1 | 2319 | UTR 3 | NP_001265140.1 | |||
NM_001278212.1 | 2319 | UTR 3 | NP_001265141.1 | |||
NM_001278213.1 | 2319 | UTR 3 | NP_001265142.1 | |||
NM_001278214.1 | 2319 | UTR 3 | NP_001265143.1 | |||
NM_018205.3 | 2319 | UTR 3 | NP_060675.1 | |||
NM_018239.3 | 2319 | UTR 3 | NP_060709.2 | |||
NM_207119.2 | 2319 | UTR 3 | NP_997002.1 | |||
XM_017016374.1 | 2319 | UTR 3 | XP_016871863.1 | |||
XM_017016375.1 | 2319 | UTR 3 | XP_016871864.1 |