Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186975 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCTE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TCTE1 - t-complex-associated-testis-expressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182539.3 | 2081 | Silent Mutation | ATA,ATC | I,I 491 | NP_872345.2 | |
XM_005248872.1 | 2081 | Silent Mutation | ATA,ATC | I,I 491 | XP_005248929.1 | |
XM_005248873.2 | 2081 | Silent Mutation | ATA,ATC | I,I 404 | XP_005248930.1 | |
XM_005248874.2 | 2081 | Silent Mutation | ATA,ATC | I,I 392 | XP_005248931.1 | |
XM_006715009.3 | 2081 | Silent Mutation | ATA,ATC | I,I 392 | XP_006715072.1 | |
XM_011514336.2 | 2081 | Silent Mutation | ATA,ATC | I,I 491 | XP_011512638.1 | |
XM_011514338.2 | 2081 | Silent Mutation | ATA,ATC | I,I 491 | XP_011512640.1 | |
XM_011514339.2 | 2081 | Silent Mutation | ATA,ATC | I,I 392 | XP_011512641.1 | |
XM_011514340.1 | 2081 | Silent Mutation | ATA,ATC | I,I 341 | XP_011512642.1 |
TMEM151B - transmembrane protein 151B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137560.1 | 2081 | Intron | NP_001131032.1 |
Set Membership: |
Validated |