Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAAATGGAGGCCCCAGAAAGAAG[C/T]TGCAAGGAAACATCTGCAGTTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603322 MIM: 609507 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFB6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
NDUFB6 - NADH:ubiquinone oxidoreductase subunit B6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199987.1 | Intron | NP_001186916.1 | ||||
NM_002493.4 | Intron | NP_002484.1 | ||||
NM_182739.2 | Intron | NP_877416.1 |
TOPORS - TOP1 binding arginine/serine rich protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOPORS-AS1 - TOPORS antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |