Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602193 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC29A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC29A1 - solute carrier family 29 member 1 (Augustine blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001078175.2 | Intron | NP_001071643.1 | ||||
NM_001078177.1 | Intron | NP_001071645.1 | ||||
NM_001304462.1 | Intron | NP_001291391.1 | ||||
NM_001304463.1 | Intron | NP_001291392.1 | ||||
NM_001304465.1 | Intron | NP_001291394.1 | ||||
NM_001304466.1 | Intron | NP_001291395.1 | ||||
XM_005248876.4 | Intron | XP_005248933.1 | ||||
XM_005248878.3 | Intron | XP_005248935.1 | ||||
XM_005248879.3 | Intron | XP_005248936.1 | ||||
XM_005248880.3 | Intron | XP_005248937.1 | ||||
XM_005248881.3 | Intron | XP_005248938.1 | ||||
XM_005248882.3 | Intron | XP_005248939.1 | ||||
XM_011514341.2 | Intron | XP_011512643.1 |
Set Membership: |
HapMap |