Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607790 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TET1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TET1 - tet methylcytosine dioxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030625.2 | 476 | UTR 5 | NP_085128.2 | |||
XM_011540204.2 | 476 | UTR 5 | XP_011538506.1 | |||
XM_011540205.2 | 476 | Intron | XP_011538507.1 | |||
XM_011540206.2 | 476 | UTR 5 | XP_011538508.1 | |||
XM_011540207.2 | 476 | Intron | XP_011538509.1 | |||
XM_017016686.1 | 476 | Intron | XP_016872175.1 | |||
XM_017016687.1 | 476 | Intron | XP_016872176.1 | |||
XM_017016688.1 | 476 | Intron | XP_016872177.1 | |||
XM_017016689.1 | 476 | Intron | XP_016872178.1 |