Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAATCAAGAGAACTGAAAACAT[C/T]GACTTTTTACAAGCACATAGAGCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154545 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MBL2 - mannose binding lectin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000242.2 | 2015 | UTR 3 | NP_000233.1 | |||
XM_006717861.3 | 2015 | UTR 3 | XP_006717924.1 | |||
XM_011539816.2 | 2015 | UTR 3 | XP_011538118.1 |
Set Membership: |
HapMap |