Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGCCACCACGCCCGGCCAAAAA[C/T]TGACTTTCTATATCGCCCCCCACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600046 MIM: 608872 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABCA1 - ATP binding cassette subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005502.3 | Intron | NP_005493.2 | ||||
XM_005251773.2 | Intron | XP_005251830.1 | ||||
XM_005251776.2 | Intron | XP_005251833.1 | ||||
XM_011518339.2 | Intron | XP_011516641.1 | ||||
XM_011518340.2 | Intron | XP_011516642.1 | ||||
XM_011518341.2 | Intron | XP_011516643.1 | ||||
XM_011518342.2 | Intron | XP_011516644.1 | ||||
XM_011518344.2 | Intron | XP_011516646.1 | ||||
XM_017014378.1 | Intron | XP_016869867.1 | ||||
XM_017014379.1 | Intron | XP_016869868.1 | ||||
XM_017014380.1 | Intron | XP_016869869.1 | ||||
XM_017014381.1 | Intron | XP_016869870.1 | ||||
XM_017014382.1 | Intron | XP_016869871.1 |
NIPSNAP3B - nipsnap homolog 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |