Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615125 MIM: 602010 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD13C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ANKRD13C - ankyrin repeat domain 13C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030816.4 | 4177 | UTR 3 | NP_110443.3 | |||
XM_005271234.3 | 4177 | UTR 3 | XP_005271291.1 | |||
XM_005271235.3 | 4177 | Intron | XP_005271292.1 | |||
XM_006710929.3 | 4177 | Intron | XP_006710992.1 | |||
XM_017002413.1 | 4177 | UTR 3 | XP_016857902.1 | |||
XM_017002414.1 | 4177 | Intron | XP_016857903.1 | |||
XM_017002415.1 | 4177 | Intron | XP_016857904.1 | |||
XM_017002416.1 | 4177 | UTR 3 | XP_016857905.1 |
SRSF11 - serine and arginine rich splicing factor 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |