Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCGGTGGCGTGGCAGGAAGGCC[A/G]TTCCTCCAGGAAAAATTCTGAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600777 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP77 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP77 - cilia and flagella associated protein 77 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282957.1 | Intron | NP_001269886.1 | ||||
NM_207417.2 | Intron | NP_997300.1 | ||||
XM_011518670.2 | Intron | XP_011516972.1 | ||||
XM_011518671.1 | Intron | XP_011516973.1 | ||||
XM_017014711.1 | Intron | XP_016870200.1 |
TTF1 - transcription termination factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |