Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGGGAGGAGTTTATTTTATGC[A/T]GTTTAAAACATATATTACATATGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602938 MIM: 611854 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAAT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BAAT - bile acid-CoA:amino acid N-acyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL50 - mitochondrial ribosomal protein L50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |