Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605933 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCSTAMP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DCSTAMP - dendrocyte expressed seven transmembrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257317.1 | Intron | NP_001244246.1 | ||||
NM_030788.3 | Intron | NP_110415.1 | ||||
XM_011517321.1 | Intron | XP_011515623.1 | ||||
XM_011517324.1 | Intron | XP_011515626.1 | ||||
XM_017013878.1 | Intron | XP_016869367.1 | ||||
XM_017013879.1 | Intron | XP_016869368.1 | ||||
XM_017013880.1 | Intron | XP_016869369.1 |