Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAACACCCTATGAAGTCTCTGTT[C/T]GTGGCCCTGAAGGCAATGCCAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604351 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR548AU PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR548AU - microRNA 548au | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHF2 - PHD finger protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005392.3 | Intron | NP_005383.3 | ||||
XM_005252051.2 | Intron | XP_005252108.1 | ||||
XM_006717143.2 | Intron | XP_006717206.1 |