Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614772 MIM: 605052 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COA6 - cytochrome c oxidase assembly factor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TARBP1 - TAR (HIV-1) RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005646.3 | Intron | NP_005637.3 | ||||
XM_005273234.3 | Intron | XP_005273291.1 | ||||
XM_011544263.2 | Intron | XP_011542565.1 | ||||
XM_017002194.1 | Intron | XP_016857683.1 | ||||
XM_017002195.1 | Intron | XP_016857684.1 | ||||
XM_017002196.1 | Intron | XP_016857685.1 | ||||
XM_017002197.1 | Intron | XP_016857686.1 |