Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCACCCCACACAGCAGGCCACC[A/G]GGGGACAGCTGTTCTCCCTTGTCCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
OXNAD1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RFTN1 - raftlin, lipid raft linker 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015150.1 | 1873 | Silent Mutation | CCC,CCT | P,P 522 | NP_055965.1 | |
XM_005264985.1 | 1873 | Silent Mutation | CCC,CCT | P,P 522 | XP_005265042.1 | |
XM_005264986.2 | 1873 | Silent Mutation | CCC,CCT | P,P 522 | XP_005265043.1 | |
XM_006713069.1 | 1873 | Silent Mutation | CCC,CCT | P,P 522 | XP_006713132.1 | |
XM_011533529.1 | 1873 | Silent Mutation | CCC,CCT | P,P 557 | XP_011531831.1 | |
XM_011533530.1 | 1873 | Silent Mutation | CCC,CCT | P,P 522 | XP_011531832.1 | |
XM_011533531.1 | 1873 | UTR 3 | XP_011531833.1 | |||
XM_017006005.1 | 1873 | Intron | XP_016861494.1 | |||
XM_017006006.1 | 1873 | Intron | XP_016861495.1 | |||
XM_017006007.1 | 1873 | UTR 3 | XP_016861496.1 |