Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGTGGGTGCACAGTTTAATAAA[C/T]GCCACCAACGGTAGCTGAGCCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153615 MIM: 615427 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CAPG - capping actin protein, gelsolin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256139.1 | Intron | NP_001243068.1 | ||||
NM_001256140.1 | Intron | NP_001243069.1 | ||||
NM_001320732.1 | Intron | NP_001307661.1 | ||||
NM_001320733.1 | Intron | NP_001307662.1 | ||||
NM_001320734.1 | Intron | NP_001307663.1 | ||||
NM_001747.3 | Intron | NP_001738.2 | ||||
XM_011533122.1 | Intron | XP_011531424.1 | ||||
XM_011533123.1 | Intron | XP_011531425.1 |
ELMOD3 - ELMO domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |