Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGGTTTTTTGTTTTTCATCTGTA[A/G]CAGGGGATAGGTTTGGCTGAGCAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615427 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELMOD3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ELMOD3 - ELMO domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135021.1 | Intron | NP_001128493.1 | ||||
NM_001135022.1 | Intron | NP_001128494.1 | ||||
NM_001135023.1 | Intron | NP_001128495.1 | ||||
NM_032213.4 | Intron | NP_115589.2 | ||||
XM_005264596.3 | Intron | XP_005264653.1 | ||||
XM_005264597.3 | Intron | XP_005264654.1 | ||||
XM_005264598.3 | Intron | XP_005264655.1 | ||||
XM_017005078.1 | Intron | XP_016860567.1 | ||||
XM_017005079.1 | Intron | XP_016860568.1 |
RETSAT - retinol saturase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |