Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601141 MIM: 606213 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNAB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KCNAB1 - potassium voltage-gated channel subfamily A member regulatory beta subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308217.1 | 2215 | Intron | NP_001295146.1 | |||
NM_001308222.1 | 2215 | Intron | NP_001295151.1 | |||
NM_003471.3 | 2215 | Intron | NP_003462.2 | |||
NM_172159.3 | 2215 | Intron | NP_751891.1 | |||
NM_172160.2 | 2215 | Intron | NP_751892.1 |
SSR3 - signal sequence receptor subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308197.1 | 2215 | UTR 3 | NP_001295126.1 | |||
NM_001308204.1 | 2215 | UTR 3 | NP_001295133.1 | |||
NM_001308205.1 | 2215 | UTR 3 | NP_001295134.1 | |||
NM_007107.4 | 2215 | UTR 3 | NP_009038.1 |