Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAAATCTAGGTGGTTAGAAGATT[C/T]TTAAATGTATAATTTTTTGGCGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC190 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC190 - coiled-coil domain containing 190 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178550.4 | 5909 | Intron | NP_848645.3 | |||
XM_005245125.3 | 5909 | UTR 3 | XP_005245182.1 | |||
XM_006711289.3 | 5909 | UTR 3 | XP_006711352.1 | |||
XM_011509477.2 | 5909 | UTR 3 | XP_011507779.1 |