Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTATTTTGCGACACACACAAAA[G/T]TCAGGGTCTTCTGAAGGTTCAGGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610890 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS7BP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RGS7BP - regulator of G-protein signaling 7 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029875.2 | Intron | NP_001025046.1 | ||||
NM_001271890.1 | Intron | NP_001258819.1 | ||||
NM_001271891.1 | Intron | NP_001258820.1 | ||||
XM_005248502.3 | Intron | XP_005248559.1 |
Set Membership: |
HapMap Validated |