Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300292 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FOXP3 - forkhead box P3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R3F - protein phosphatase 1 regulatory subunit 3F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184745.1 | 492 | Intron | NP_001171674.1 | |||
NM_033215.4 | 492 | Intron | NP_149992.3 | |||
XM_005272687.3 | 492 | Intron | XP_005272744.1 | |||
XM_017029932.1 | 492 | UTR 5 | XP_016885421.1 | |||
XM_017029933.1 | 492 | UTR 5 | XP_016885422.1 | |||
XM_017029934.1 | 492 | Intron | XP_016885423.1 | |||
XM_017029935.1 | 492 | Intron | XP_016885424.1 |
Set Membership: |
HapMap Validated |