Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610523 MIM: 114850 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP41 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CEP41 - centrosomal protein 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257158.1 | 5445 | UTR 3 | NP_001244087.1 | |||
NM_001257159.1 | 5445 | UTR 3 | NP_001244088.1 | |||
NM_001257160.1 | 5445 | Intron | NP_001244089.1 | |||
NM_018718.2 | 5445 | UTR 3 | NP_061188.1 | |||
XM_011516709.2 | 5445 | Intron | XP_011515011.1 | |||
XM_011516710.2 | 5445 | Intron | XP_011515012.1 |
CPA1 - carboxypeptidase A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |