Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACACGTGGGTCCTCAGTTTCAG[C/G]GAGCTACACAGGTACAGGGGAAGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615720 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928583 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101928583 - uncharacterized LOC101928583 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A14 - solute carrier family 7 member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020949.2 | 7893 | UTR 3 | NP_066000.2 |
Set Membership: |
HapMap |