Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCAGCACCCCAGTGTTGAGTGC[A/C]TTGACCTGCAGAGGGGAGCCTGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608237 MIM: 608527 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAL3ST2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GAL3ST2 - galactose-3-O-sulfotransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107986004 - uncharacterized LOC107986004 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEU4 - neuraminidase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167599.2 | 281 | UTR 5 | NP_001161071.1 | |||
NM_001167600.2 | 281 | Intron | NP_001161072.1 | |||
NM_001167601.2 | 281 | Intron | NP_001161073.1 | |||
NM_001167602.2 | 281 | UTR 5 | NP_001161074.1 | |||
NM_080741.3 | 281 | UTR 5 | NP_542779.2 |
Set Membership: |
HapMap Validated |