Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147781 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL4R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL4R - interleukin 4 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000418.3 | Intron | NP_000409.1 | ||||
NM_001257406.1 | Intron | NP_001244335.1 | ||||
NM_001257407.1 | Intron | NP_001244336.1 | ||||
NM_001257997.1 | Intron | NP_001244926.1 | ||||
XM_005255308.2 | Intron | XP_005255365.1 | ||||
XM_006721043.1 | Intron | XP_006721106.1 | ||||
XM_011545825.1 | Intron | XP_011544127.1 | ||||
XM_011545826.2 | Intron | XP_011544128.1 | ||||
XM_011545827.2 | Intron | XP_011544129.1 | ||||
XM_011545828.2 | Intron | XP_011544130.1 | ||||
XM_011545829.1 | Intron | XP_011544131.1 | ||||
XM_011545830.2 | Intron | XP_011544132.1 | ||||
XM_011545831.1 | Intron | XP_011544133.1 | ||||
XM_011545832.1 | Intron | XP_011544134.1 | ||||
XM_011545833.2 | Intron | XP_011544135.1 | ||||
XM_011545834.2 | Intron | XP_011544136.1 | ||||
XM_017023211.1 | Intron | XP_016878700.1 |