Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC74A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRRC74A - leucine rich repeat containing 74A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194287.2 | 314 | Intron | NP_919263.2 | |||
XM_011536479.1 | 314 | Intron | XP_011534781.1 | |||
XM_011536483.1 | 314 | Intron | XP_011534785.1 | |||
XM_011536487.1 | 314 | Intron | XP_011534789.1 | |||
XM_017021032.1 | 314 | Intron | XP_016876521.1 | |||
XM_017021033.1 | 314 | Intron | XP_016876522.1 | |||
XM_017021034.1 | 314 | Intron | XP_016876523.1 | |||
XM_017021035.1 | 314 | Intron | XP_016876524.1 | |||
XM_017021036.1 | 314 | Intron | XP_016876525.1 | |||
XM_017021037.1 | 314 | UTR 5 | XP_016876526.1 | |||
XM_017021038.1 | 314 | Intron | XP_016876527.1 | |||
XM_017021039.1 | 314 | Intron | XP_016876528.1 | |||
XM_017021040.1 | 314 | Intron | XP_016876529.1 | |||
XM_017021041.1 | 314 | Intron | XP_016876530.1 |