Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCAAGAGACCTCCCCCTCCCCCC[A/C]AAAAAAAACTTTCTAACTGGATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608663 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANO6 - anoctamin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025356.2 | Intron | NP_001020527.2 | ||||
NM_001142678.1 | Intron | NP_001136150.1 | ||||
NM_001142679.1 | Intron | NP_001136151.1 | ||||
NM_001204803.1 | Intron | NP_001191732.1 | ||||
XM_005268706.4 | Intron | XP_005268763.1 | ||||
XM_005268707.3 | Intron | XP_005268764.1 |
PLEKHA8P1 - pleckstrin homology domain containing A8 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |