Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108731 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP2B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP2B1 - ATPase plasma membrane Ca2+ transporting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001323.1 | Intron | NP_001001323.1 | ||||
NM_001682.2 | Intron | NP_001673.2 | ||||
XM_011538407.2 | Intron | XP_011536709.1 | ||||
XM_017019357.1 | Intron | XP_016874846.1 | ||||
XM_017019358.1 | Intron | XP_016874847.1 | ||||
XM_017019359.1 | Intron | XP_016874848.1 | ||||
XM_017019360.1 | Intron | XP_016874849.1 |
Set Membership: |
HapMap |