Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605874 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNK9 - potassium two pore domain channel subfamily K member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282534.1 | Intron | NP_001269463.1 | ||||
XM_011517101.2 | Intron | XP_011515403.1 | ||||
XM_011517102.2 | Intron | XP_011515404.1 | ||||
XM_011517103.2 | Intron | XP_011515405.1 | ||||
XM_017013530.1 | Intron | XP_016869019.1 | ||||
XM_017013531.1 | Intron | XP_016869020.1 |