Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616790 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP4R4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP4R4 - protein phosphatase 4 regulatory subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020958.2 | Intron | NP_066009.2 | ||||
NM_058237.1 | Intron | NP_478144.1 | ||||
XM_005267930.2 | Intron | XP_005267987.1 | ||||
XM_011537038.2 | Intron | XP_011535340.1 | ||||
XM_011537039.2 | Intron | XP_011535341.1 | ||||
XM_011537040.2 | Intron | XP_011535342.1 | ||||
XM_017021528.1 | Intron | XP_016877017.1 | ||||
XM_017021529.1 | Intron | XP_016877018.1 | ||||
XM_017021530.1 | Intron | XP_016877019.1 |
Set Membership: |
HapMap |