Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTCATAGAAATAGTGGGGGCTT[A/T]ATGTTTTCCCATAATATTTTGGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM221A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM221A - family with sequence similarity 221 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127364.2 | Intron | NP_001120836.1 | ||||
NM_001127365.2 | Intron | NP_001120837.1 | ||||
NM_001300932.1 | Intron | NP_001287861.1 | ||||
NM_199136.4 | Intron | NP_954587.2 | ||||
XM_011515369.2 | Intron | XP_011513671.1 | ||||
XM_011515370.2 | Intron | XP_011513672.1 | ||||
XM_011515371.2 | Intron | XP_011513673.1 | ||||
XM_011515372.2 | Intron | XP_011513674.1 | ||||
XM_017012134.1 | Intron | XP_016867623.1 | ||||
XM_017012135.1 | Intron | XP_016867624.1 |