Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGCCATAAAAGAGTGTGAATGG[C/T]TCCCTGCAGAACTGCCACCACTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VASH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VASH1 - vasohibin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014909.4 | Intron | NP_055724.1 | ||||
XM_017021087.1 | Intron | XP_016876576.1 | ||||
XM_017021088.1 | Intron | XP_016876577.1 | ||||
XM_017021089.1 | Intron | XP_016876578.1 | ||||
XM_017021090.1 | Intron | XP_016876579.1 | ||||
XM_017021091.1 | Intron | XP_016876580.1 |