Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 311550 MIM: 314370 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CDK16 - cyclin dependent kinase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170460.1 | Intron | NP_001163931.1 | ||||
NM_006201.4 | Intron | NP_006192.1 | ||||
NM_033018.3 | Intron | NP_148978.2 | ||||
XM_011543920.2 | Intron | XP_011542222.1 | ||||
XM_011543921.1 | Intron | XP_011542223.1 | ||||
XM_011543922.1 | Intron | XP_011542224.1 | ||||
XM_011543923.1 | Intron | XP_011542225.1 | ||||
XM_011543924.1 | Intron | XP_011542226.1 | ||||
XM_011543925.2 | Intron | XP_011542227.1 | ||||
XM_011543926.1 | Intron | XP_011542228.1 | ||||
XM_011543927.2 | Intron | XP_011542229.1 | ||||
XM_011543928.1 | Intron | XP_011542230.1 | ||||
XM_017029569.1 | Intron | XP_016885058.1 | ||||
XM_017029570.1 | Intron | XP_016885059.1 | ||||
XM_017029571.1 | Intron | XP_016885060.1 | ||||
XM_017029572.1 | Intron | XP_016885061.1 | ||||
XM_017029573.1 | Intron | XP_016885062.1 |
UBA1 - ubiquitin like modifier activating enzyme 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |