Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACATCTGGAGTACTTTGAAGAAG[C/T]CGTGGACTTTATGCTGCAGCATCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614267 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ACOT6 - acyl-CoA thioesterase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037162.1 | 321 | UTR 5 | NP_001032239.1 | |||
XM_011537076.2 | 321 | Missense Mutation | GCC,GTC | A,V 249 | XP_011535378.1 |
HEATR4 - HEAT repeat containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001220484.1 | 321 | Intron | NP_001207413.1 | |||
NM_203309.2 | 321 | Intron | NP_976054.2 | |||
XM_006720143.2 | 321 | Intron | XP_006720206.1 | |||
XM_011536760.2 | 321 | Intron | XP_011535062.1 | |||
XM_011536761.2 | 321 | Intron | XP_011535063.1 | |||
XM_017021289.1 | 321 | Intron | XP_016876778.1 |
Set Membership: |
HapMap Validated |