Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614313 MIM: 611919 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ACOT1 - acyl-CoA thioesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037161.1 | 3377 | Intron | NP_001032238.1 | |||
XM_017021589.1 | 3377 | Intron | XP_016877078.1 | |||
XM_017021590.1 | 3377 | Intron | XP_016877079.1 | |||
XM_017021591.1 | 3377 | Intron | XP_016877080.1 | |||
XM_017021592.1 | 3377 | Intron | XP_016877081.1 |
C14orf169 - chromosome 14 open reading frame 169 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HEATR4 - HEAT repeat containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001220484.1 | 3377 | Silent Mutation | AAA,AAG | K,K 849 | NP_001207413.1 | |
NM_203309.2 | 3377 | Silent Mutation | AAA,AAG | K,K 849 | NP_976054.2 | |
XM_006720143.2 | 3377 | Silent Mutation | AAA,AAG | K,K 849 | XP_006720206.1 | |
XM_011536760.2 | 3377 | Silent Mutation | AAA,AAG | K,K 849 | XP_011535062.1 | |
XM_011536761.2 | 3377 | Intron | XP_011535063.1 | |||
XM_017021289.1 | 3377 | Silent Mutation | AAA,AAG | K,K 849 | XP_016876778.1 |
Set Membership: |
HapMap Validated |