Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT52 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IFT52 - intraflagellar transport 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303458.2 | 582 | Silent Mutation | GTA,GTG | V,V 53 | NP_001290387.1 | |
NM_001303459.2 | 582 | Silent Mutation | GTA,GTG | V,V 53 | NP_001290388.1 | |
NM_001323578.1 | 582 | UTR 5 | NP_001310507.1 | |||
NM_001323579.1 | 582 | UTR 5 | NP_001310508.1 | |||
NM_001323580.1 | 582 | UTR 5 | NP_001310509.1 | |||
NM_001323581.1 | 582 | UTR 5 | NP_001310510.1 | |||
NM_016004.4 | 582 | Silent Mutation | GTA,GTG | V,V 53 | NP_057088.2 | |
XM_017027863.1 | 582 | UTR 5 | XP_016883352.1 |