Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607589 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT52 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IFT52 - intraflagellar transport 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303458.2 | Intron | NP_001290387.1 | ||||
NM_001303459.2 | Intron | NP_001290388.1 | ||||
NM_001323578.1 | Intron | NP_001310507.1 | ||||
NM_001323579.1 | Intron | NP_001310508.1 | ||||
NM_001323580.1 | Intron | NP_001310509.1 | ||||
NM_001323581.1 | Intron | NP_001310510.1 | ||||
NM_016004.4 | Intron | NP_057088.2 | ||||
XM_017027863.1 | Intron | XP_016883352.1 |
SGK2 - SGK2, serine/threonine kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |