Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAAGTATTGAGCTCGCCATTTTT[A/G]TGGGTCTTCCTCTACCCCCAACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601714 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEAD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TEAD4 - TEA domain transcription factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003213.3 | Intron | NP_003204.2 | ||||
NM_201441.2 | Intron | NP_958849.1 | ||||
NM_201443.2 | Intron | NP_958851.1 |
Set Membership: |
Validated |