Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609740 MIM: 601711 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF19 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PHF19 - PHD finger protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009936.2 | 3955 | Intron | NP_001009936.1 | |||
NM_001286840.1 | 3955 | Intron | NP_001273769.1 | |||
NM_001286842.1 | 3955 | Intron | NP_001273771.1 | |||
NM_001286843.1 | 3955 | Intron | NP_001273772.1 | |||
NM_015651.2 | 3955 | Intron | NP_056466.1 | |||
XM_005251906.2 | 3955 | Intron | XP_005251963.1 | |||
XM_011518509.2 | 3955 | Intron | XP_011516811.1 | |||
XM_011518511.2 | 3955 | Intron | XP_011516813.1 | |||
XM_011518515.2 | 3955 | Intron | XP_011516817.1 | |||
XM_011518516.2 | 3955 | Intron | XP_011516818.1 | |||
XM_017014612.1 | 3955 | Intron | XP_016870101.1 | |||
XM_017014613.1 | 3955 | Intron | XP_016870102.1 |
TRAF1 - TNF receptor associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190945.1 | 3955 | UTR 3 | NP_001177874.1 | |||
NM_001190947.1 | 3955 | UTR 3 | NP_001177876.1 | |||
NM_005658.4 | 3955 | UTR 3 | NP_005649.1 |
Set Membership: |
HapMap |