Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAACTTGACAAGTCACATTTAATA[C/T]AGATTCTGACACAGAATAATAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611524 MIM: 605634 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RARS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RARS2 - arginyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318785.1 | 2392 | UTR 3 | NP_001305714.1 | |||
NM_020320.4 | 2392 | UTR 3 | NP_064716.2 | |||
XM_011535948.2 | 2392 | UTR 3 | XP_011534250.1 | |||
XM_011535949.2 | 2392 | Intron | XP_011534251.1 | |||
XM_017011072.1 | 2392 | UTR 3 | XP_016866561.1 | |||
XM_017011073.1 | 2392 | UTR 3 | XP_016866562.1 | |||
XM_017011074.1 | 2392 | UTR 3 | XP_016866563.1 | |||
XM_017011075.1 | 2392 | UTR 3 | XP_016866564.1 | |||
XM_017011076.1 | 2392 | UTR 3 | XP_016866565.1 | |||
XM_017011077.1 | 2392 | UTR 3 | XP_016866566.1 | |||
XM_017011078.1 | 2392 | Intron | XP_016866567.1 | |||
XM_017011079.1 | 2392 | UTR 3 | XP_016866568.1 | |||
XM_017011080.1 | 2392 | Intron | XP_016866569.1 | |||
XM_017011081.1 | 2392 | Intron | XP_016866570.1 |
SLC35A1 - solute carrier family 35 member A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |