Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611299 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
G2E3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
G2E3 - G2/M-phase specific E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCFD1 - sec1 family domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257376.1 | Intron | NP_001244305.1 | ||||
NM_001283031.1 | Intron | NP_001269960.1 | ||||
NM_001283032.1 | Intron | NP_001269961.1 | ||||
NM_001283033.1 | Intron | NP_001269962.1 | ||||
NM_016106.3 | Intron | NP_057190.2 | ||||
NM_182835.2 | Intron | NP_878255.1 | ||||
XM_005267469.2 | Intron | XP_005267526.1 | ||||
XM_011536586.2 | Intron | XP_011534888.1 | ||||
XM_011536587.2 | Intron | XP_011534889.1 | ||||
XM_011536588.2 | Intron | XP_011534890.1 | ||||
XM_011536589.1 | Intron | XP_011534891.1 | ||||
XM_017021113.1 | Intron | XP_016876602.1 |
Set Membership: |
Validated |