Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602884 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMDS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GMDS - GDP-mannose 4,6-dehydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253846.1 | Intron | NP_001240775.1 | ||||
NM_001500.3 | Intron | NP_001491.1 | ||||
XM_006715066.2 | Intron | XP_006715129.1 | ||||
XM_011514500.1 | Intron | XP_011512802.1 | ||||
XM_011514502.2 | Intron | XP_011512804.1 | ||||
XM_011514503.2 | Intron | XP_011512805.1 | ||||
XM_011514505.1 | Intron | XP_011512807.1 | ||||
XM_011514506.1 | Intron | XP_011512808.1 | ||||
XM_011514507.1 | Intron | XP_011512809.1 | ||||
XM_017010752.1 | Intron | XP_016866241.1 |
Set Membership: |
HapMap Validated |