Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613947 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPATA6 - spermatogenesis associated 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286238.1 | 4053 | Intron | NP_001273167.1 | |||
NM_001286239.1 | 4053 | Intron | NP_001273168.1 | |||
NM_019073.3 | 4053 | Intron | NP_061946.1 | |||
XM_006710699.2 | 4053 | UTR 3 | XP_006710762.1 | |||
XM_006710700.2 | 4053 | Intron | XP_006710763.1 | |||
XM_006710701.3 | 4053 | Intron | XP_006710764.1 | |||
XM_011541606.1 | 4053 | Intron | XP_011539908.1 | |||
XM_011541607.2 | 4053 | Intron | XP_011539909.1 | |||
XM_011541608.1 | 4053 | Intron | XP_011539910.1 | |||
XM_011541609.2 | 4053 | Intron | XP_011539911.1 | |||
XM_017001509.1 | 4053 | Intron | XP_016856998.1 | |||
XM_017001510.1 | 4053 | Intron | XP_016856999.1 | |||
XM_017001511.1 | 4053 | Intron | XP_016857000.1 | |||
XM_017001512.1 | 4053 | Intron | XP_016857001.1 | |||
XM_017001513.1 | 4053 | Intron | XP_016857002.1 | |||
XM_017001514.1 | 4053 | Intron | XP_016857003.1 | |||
XM_017001515.1 | 4053 | Intron | XP_016857004.1 |