Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614716 MIM: 605688 MIM: 162080 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARMIL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CARMIL3 - capping protein regulator and myosin 1 linker 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CPNE6 - copine 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRL - neural retina leucine zipper | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006177.3 | 2507 | Intron | NP_006168.1 | |||
XM_005267708.4 | 2507 | UTR 3 | XP_005267765.1 | |||
XM_005267709.3 | 2507 | UTR 3 | XP_005267766.1 | |||
XM_005267710.3 | 2507 | UTR 3 | XP_005267767.1 | |||
XM_011536801.2 | 2507 | UTR 3 | XP_011535103.2 | |||
XM_011536802.1 | 2507 | UTR 3 | XP_011535104.1 | |||
XM_011536804.2 | 2507 | UTR 3 | XP_011535106.1 | |||
XM_011536805.2 | 2507 | UTR 3 | XP_011535107.1 | |||
XM_011536806.2 | 2507 | UTR 3 | XP_011535108.2 | |||
XM_017021350.1 | 2507 | UTR 3 | XP_016876839.1 |
Set Membership: |
HapMap |