Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCACACTGTAAAGTTAGACTTT[G/T]TCATATGACTTCCTTGCGGCAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603449 MIM: 609213 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUVBL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RUVBL1 - RuvB like AAA ATPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319084.1 | Intron | NP_001306013.1 | ||||
NM_001319086.1 | Intron | NP_001306015.1 | ||||
NM_003707.2 | Intron | NP_003698.1 | ||||
XM_005247841.3 | Intron | XP_005247898.1 | ||||
XM_011513248.1 | Intron | XP_011511550.1 | ||||
XM_011513249.2 | Intron | XP_011511551.1 | ||||
XM_017007356.1 | Intron | XP_016862845.1 | ||||
XM_017007357.1 | Intron | XP_016862846.1 |
RUVBL1-AS1 - RUVBL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEC61A1 - Sec61 translocon alpha 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013336.3 | Intron | NP_037468.1 |
Set Membership: |
Validated |